Electronic Hand-Held Crimpers and Decrimpers, Level 3 High Performance Applications
Electronic Hand-Held Crimpers and Decrimpers, Level 3 High Performance Applications
Actual product may vary
Electronic Hand-Held Crimpers and Decrimpers, Level 3 High Performance Applications
Electronic Hand-Held Crimpers and Decrimpers, Level 3 High Performance Applications
Electronic Hand-Held Crimpers and Decrimpers, Level 3 High Performance Applications
Electronic Hand-Held Crimpers and Decrimpers, Level 3 High Performance Applications
Electronic Hand-Held Crimpers and Decrimpers, Level 3 High Performance Applications
Electronic Hand-Held Crimpers and Decrimpers, Level 3 High Performance Applications
Use Thermo Scientific™ Electronic Hand-Held Vial Crimpers and Decrimpers to attach or remove crimp caps from vials, respectively.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeSize
60180-EDCB13Electronic Decrimper13 mm
60180-ECR11Electronic Crimper11 mm
60180-ECR13Electronic Crimper13 mm
60180-ECR20Electronic Crimper20 mm
60180-ECR8Electronic Crimper8 mm
60180-EDCB11Electronic Decrimper11 mm
60180-EDCB20Electronic Decrimper20 mm
60180-ECRHBPower Supply110/240 V
Catalog number 60180-EDCB13
HSN 84672900
Price (INR)
157,736.00
Each
Add to cart
Type:
Electronic Decrimper
Size:
13 mm
Price (INR)
157,736.00
Each
Add to cart
Use Thermo Scientific™ Electronic Hand-Held Vial Crimpers and Decrimpers to attach or remove crimp caps from vials, respectively. These hand-held, high-power crimpers and decrimpers are available in 8 mm, 11 mm, 13 mm, and 20 mm sizes and allow you to seal or remove crimp caps with a simple, one-handed push of a button. Use for crimping of vials with aluminum, pressure-release, and bi-metal caps.
The electronic crimper's message window conveniently provides all-important information at one glance. This device is Performance Level 3, which means it is designed to protect your most valuable samples and deliver consistent, reproducible results for your most demanding analyses.
Features
Hand-Held Crimper and Decrimper
  • The battery charge icon on the color screen changes to red when battery is low
  • Includes a fully rechargeable lithium ion battery and a universal power supply/recharger with international plug adapters
The Message Window Displays…
  • The actual crimp power
  • An error message after an incorrect crimping process
  • Statistics and log of recent crimping results
  • In the language of your choice (English, Chinese, Japanese, German, French, Spanish, Italian, Korean)
These Electronic Vial Crimpers also feature adjustable settings for best crimp results and a motor with extended lifetime and no particle emission.
Specifications
OperationManual via Push Buttons
Power SupplyFully Rechargeable Lithium Ion Battery
TypeElectronic Decrimper
For Use With (Equipment)13 mm Crimp Vials
Size13 mm
Product LineSureSTART Level 3
QuantityEach
Unit SizeEach