Gas Chromatography 3-Port SilFlow Device
Product Image
Gas Chromatography 3-Port SilFlow Device
Switch your GC without the need to vent using the Thermo Scientific™ 3-Port SilFlow™ Device.
Have Questions?
Catalog NumberQuantity
60201-3981/Ea.
Catalog number 60201-398
HSN 73269099
Price (INR)
92,557.00
Each
Add to cart
Quantity:
1/Ea.
Price (INR)
92,557.00
Each
Add to cart
Switch your GC without the need to vent using the Thermo Scientific™ 3-Port SilFlow™ Device. This device comprises a gas module with microfluidics and Thermo Scientific™ SilTite™ FingerTite fittings for easy set up and a reliable seal.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Type3-Port SilFlow MCD
Description3-Port SilFlow MCD (0.25/0.32)
Quantity1/Ea.
For Use With (Equipment)Gas Chromatography Systems
Unit SizeEach