HyperSep™ SPE Plates
Thermo Scientific™
HyperSep™ SPE Plates
Innovative HyperSep™ Solid Phase Extraction Plates deliver clean, highly reproducible sample extracts with lower elution volumes—for greater sensitivity, reliability and cost savings.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludes
60300-10-2-96-WAX20 mg/1 mL 96-well plate
60300-10-1-96-WAX10 mg/1 mL 96-well plate
60300-03-1-96-WAX3 mg/0.6 mL 96-well plate
Catalog number 60300-10-2-96-WAX
Price (INR)
-
Includes:
20 mg/1 mL 96-well plate
Obtain clean, highly reproducible sample extracts with lower elution volumes using Thermo Scientific™ HyperSep™ Solid Phase Extraction Plates. The enhanced performance gives you greater confidence in your analytical results and lowers operating costs—without compromising ease of use or requiring complex method development. Thanks to an innovative and unique frit-less design, HyperSep SPE Well Plates deliver unparalleled performance, including:
  • Higher levels of reproducibility
  • Lower elution volumes
  • High extract cleanliness
HyperSep Solid Phase Extraction Plates achieve excellent recovery levels even with low volumes of extract solvents, resulting in a more concentrated analyte and increased sensitivity. You'll also realize additional cost and time saving from faster sample dry-down time and reduced solvent usage.
  • 10mg/2mL 96 well plate format
  • Hydrophobic Reversed Phase (HRP), Strong Cation Exchanger (SCX) and Strong Anion Exchanger (SAX) phase types
  • HyperSep products are also available in cartridge format.
Specifications
For Use With (Application)Bioanalytical Applications - Weak Ion-Exchange Retention of Strong Acidic Compounds. Sorbent Charge Can be Activated or Deactivated. Complementary Reversed Phase Retention of Neutral Compounds
Includes20 mg/1 mL 96-well plate
Bed Weight10 mg
Product LineHyperSep
Type96-well Plate
Volume (Metric) Well2 mL
Unit SizeEach