HyperSep™ Retain PEP Plates
HyperSep™ Retain PEP Plates
Actual product may vary

HyperSep™ Retain PEP Plates

Get balanced retention for polar and nonpolar analytes with HyperSep Retain PEP Plates.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberBed WeightVolume (Metric)
60303-20730 mg1 mL
60306-20730 mg2 mL
60306-20860 mg2 mL
Catalog number 60303-207
HSN 90279090
Price (INR)
51,235.00
Each
Add to cart
Bed Weight:
30 mg
Volume (Metric):
1 mL
Price (INR)
51,235.00
Each
Add to cart
Get balanced retention for polar and nonpolar analytes with Thermo Scientific™ HyperSep™ Retain PEP Plates, packed with a high capacity, high purity, highly porous polystyrene DVB material modified with urea functional groups. With chemistry pH stable 0 to 14, the of these cartridges in SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.
Features: Ideal for large samples, Hypersep Retain SPE wells conform to industry standard configurations and allow efficient routine processing by vacuum, positive pressure or centrifugation.
  • [bullet] High, consistent recoveries
  • [bullet] Fast and easy sample preparation and method development
  • [bullet] Reproducible, reliable sample preparation in a traditional format
  • [bullet] Available in a wide range of chemistries and bed weights
Applications:
  • [bullet] Drugs and metabolites in biological fluids
  • [bullet] Peptides in serum, plasma or biological fluids
  • [bullet] Environmental samples
For Use With: The Thermo Scientific™ HyperSep-96™ System
Specifications
DescriptionRetain PEP 30 mg/1 mL 96 Removable Well Plate, 1 pk
Bed Weight30 mg
For Use With (Application)Retention of Polar and Nonpolar Analytes
Particle Size30 to 50 μm
Product LineHyperSep
SorbentHyperSep Urea-Modified Polystyrene DVB
TypeSolid Phase Extraction Well Plate
Volume (Metric)1 mL
Volume (Metric) Well1 mL
Quantity1 Pack
Unit SizeEach