Product Image
Applied Biosystems™

ASSY, Sample Collector .50LD Z

Have Questions?
Catalog NumberQuantity
6035951 unit
Catalog number 603595
Price (INR)
-
Quantity:
1 unit
This is a spare part for the 310 Genetic Analyzer.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)310 Genetic Analyzer
Product TypeSample Collector Assembly
Quantity1 unit
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Store at room temperature.