Viper™ Capillary Kits for Application-Specific Vanquish™ LC Systems
Thermo Scientific™
Viper™ Capillary Kits for Application-Specific Vanquish™ LC Systems
Boost your productivity and confidence, and accelerate method development using application-specific LC workflows.
Have Questions?
Catalog NumberTypeFor Use With (Equipment)For Use With (Application)
6036.0100Extension KitVanquish Flex and Horizon UHPLC, Core HPLC SystemsAutomated Method Scouting
Catalog number 6036.0100
Price (INR)
-
Type:
Extension Kit
For Use With (Equipment):
Vanquish Flex and Horizon UHPLC, Core HPLC Systems
For Use With (Application):
Automated Method Scouting
Improve chromatographic resolution by minimizing dead volume with the Thermo Scientific™ Viper™ Fingertight Fitting Systems. These pre-assembled kits allow you to quickly connect LC modules, valves, and columns for leak-free fluidic HPLC and UHPLC connections. Ideally suited for running application-specific workflows with your HPLC and LC-MS systems, like the Thermo Scientific™ Vanquish™ Method Development LC Systems, Thermo Scientific Vanquish Online 2D-LC Systems, or Thermo Scientific Vanquish Duo UHPLC Systems.
See Viper and nanoViper resources
Enjoy robust and reliable fingertight fitting LC connections. Tailored to your advanced workflow requirements, Viper application-specific capillary kits offer:
  • Virtually zero-dead volume fingertight connections
  • Easy-to-install, tool-free reusable capillaries
  • Reliable, reproducible LC connections that deliver low carryover and simplify method transfer
  • Support for a wide range of applications with different materials and operating pressures up to 1517 bar (22,000 psi)
Specifications
For Use With (Application)Automated Method Scouting
For Use With (Equipment)Vanquish Flex and Horizon UHPLC, Core HPLC Systems
TypeExtension Kit
Unit SizeEach