Product Image
Thermo Scientific™
UltiMate™ 3000 Pump Pistons
Order replacement pistons for your Thermo Scientific™ UltiMate™ 3000 LC Pump when needed.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Material
6040.0042Dionex UltiMate 3000 SD Pump, Dionex UltiMate 3000 RS Pump, Dionex UltiMate 3000 BM PumpSapphire
6040.0842Ultimate 3000 BX PumpCeramic
6043.0169Ultimate 3000 XRS PumpsSapphire
Catalog number 6040.0042
Price (INR)
-
For Use With (Equipment):
Dionex UltiMate 3000 SD Pump, Dionex UltiMate 3000 RS Pump, Dionex UltiMate 3000 BM Pump
Material:
Sapphire
Order these replacement Thermo Scientific™ Dionex™ UltiMate™ 3000 Pump Pistons when needed. Pistons are available in ceramic and sapphire to meet the needs of your specific pump.
Specifications
For Use With (Equipment)Dionex UltiMate 3000 SD Pump, Dionex UltiMate 3000 RS Pump, Dionex UltiMate 3000 BM Pump
MaterialSapphire
TypePump Piston
Unit SizeEach