UltiMate™ 3000 Pump Normal-Phase Kits
Product Image
Thermo Scientific™
UltiMate™ 3000 Pump Normal-Phase Kits
Use Thermo Scientific™ Dionex™ UltiMate™ 3000 Pump Normal-Phase Kits to adapt your LC system for normal-phase chromatography.
Have Questions?
Catalog NumberFor Use With (Equipment)
6040.1975Dionex UltiMate 3000 HPG-3200 BX Pump
Catalog number 6040.1975
Price (INR)
-
Request A Quote
For Use With (Equipment):
Dionex UltiMate 3000 HPG-3200 BX Pump
Use Thermo Scientific™ Dionex™ UltiMate™ 3000 Pump Normal Phase Kits to adapt your LC system pump for normal-phase chromatography. These kits allow you to run samples on columns featuring polar stationary phases such as those containing cyano or amino functional groups.
Specifications
For Use With (Equipment)Dionex UltiMate 3000 HPG-3200 BX Pump
TypeNormal-Phase Kit
Unit SizeEach