Viper™ Capillary Kits for LC Pumps
Viper™  Capillary Kits for LC Pumps
Actual product may vary
Viper™  Capillary Kits for LC Pumps
Viper™  Capillary Kits for LC Pumps
Thermo Scientific™
Viper™ Capillary Kits for LC Pumps
Order this single kit for all the easy, zero-dead-volume Thermo Scientific™ Viper™ Fingertight Fittings assembled with capillaries for your LC pumps.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Diameter (Metric) Outer
6040.2301ISO, LPG, or DGP Pumps for RS Systems130 μm/180 μm
6040.2302ISO, LPG, or DGP Pumps for SD Systems180 μm
6040.2308HPG Pumps for RS Systems130 μm/180 μm
6040.2309HPG Pumps for SD Systems180 μm
Catalog number 6040.2301
Price (INR)
-
Request A Quote
For Use With (Equipment):
ISO, LPG, or DGP Pumps for RS Systems
Diameter (Metric) Outer:
130 μm/180 μm
Avoid data-destroying dead volumes in the capillary connections of your HPLC or UHPLC pump connections by using Thermo Scientific™ Viper™ Capillary Kits for LC Pumps. This kit provides worry-free connections for Thermo Scientific™ Dionex™ UltiMate™ 3000 ISO, LPG, or DGP Pumps in UltiMate RSLC and SD systems. Even with conventional columns and moderate pressures, poor connections can cause retention time shifts and peak distortions. The kit contains ViperFingertight Fittings which seal at the tip of the tubing and eliminate dead volumes and make it easier to plumb your HPLC or UHPLC system.
See Viper and nanoViper resources
Worry-Free Connections
You won’t have to worry about capillary slippage or improper ferrule positioning anymore. Consistently perfect connections ensure robust chromatography with minimal interruption.
Pump-Specific Kits
  • UltiMate 3000 ISO, LPG, or DGP Pumps used in UltiMate 3000 SD Systems
  • UltiMate 3000 HPG Pumps used in UltiMate 3000 SD Systems
  • UltiMate 3000 ISO, LPG, or DGP Pumps used in UltiMate 3000 RS Systems
  • UltiMate 3000 HPG Pumps forUltiMate 3000 RS Systems
Convenient Single Kit
Stainless-steel Viper capillaries (I.D. x L, one each):
  • 0.18mm/0.007 in. x 450mm
  • 0.13mm/0.005 in. x 350mm
  • 0.13mm/0.005 in. x 250mm
Specifications
Diameter (Metric) Outer130 μm/180 μm
For Use With (Application)HPLC, UHPLC
Length (Metric)Various
MaterialStainless Steel
Max. Pressure18125 psi (1250 bar)
For Use With (Equipment)ISO, LPG, or DGP Pumps for RS Systems
Product LineViper
TypeCapillary Kit with Single System Connections
Unit SizeEach