Flow Selectors for UltiMate™ NCS-3x00RS Nano/Capillary Pumps
Actual product may vary
Thermo Scientific™
Flow Selectors for UltiMate™ NCS-3x00RS Nano/Capillary Pumps
Flow Selectors for UltiMate NCS-3x00RS Nano/Capillary Pumps are precalibrated to support given flow rate ranges for the UltiMate 3000 RSLCnano LC System.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
6041.0002NCS-3500 RS and NCP-3200 RS, Nano LC
6041.0003NCS-3500 RS and NCP-3200 RS, Capillary LC
6041.0014NCS-3500 RS and NCP-3200 RS, Micro LC
Catalog number 6041.0002
Price (INR)
-
For Use With (Equipment):
NCS-3500 RS and NCP-3200 RS, Nano LC
Choose Thermo Scientific™ Dionex™ Flow Selectors for Thermo Scientific™ UltiMate™ NCS-3x00RS Nano/Capillary Pumps that are precalibrated to support given flow rate ranges for the UltiMate 3000 RSLCnano LC System.
  • Easily exchangeable flow selector for maximum pump performance and system flexibility
Flow Selectors (recommended [settable]):
  • Nano: 50nL/min. to 1000nL/min. [1nL/min. to 1500nL/min.]
  • Capillary: 500nL/min. to 10μL/min. [1nL/min. to 15μL/min.]
  • Micro: 2.5μL/min. to 50μL/min. [1nL/min–50μL/min.]
Specifications
DescriptionFlow Selector for NCS-3500 RS and NCP-3200 RS, for Nano LC with flow range 50-1000 nL/min
For Use With (Equipment)NCS-3500 RS and NCP-3200 RS, Nano LC
Min. Flow Rate50 nL/min.
Max. Flow Rate1000 nL/min.
TypeFlow Selector
Unit SizeEach