Manual Sample Injector Valves and Mounting Kit for UltiMate™ 3000 Series Pumps
Manual Sample Injector Valves and Mounting Kit for UltiMate™ 3000 Series Pumps
Actual product may vary
Thermo Scientific™
Manual Sample Injector Valves and Mounting Kit for UltiMate™ 3000 Series Pumps
The Manual Sample Injector Valves and Mounting Kit for UltiMate™ 3000 Series Pumps include maintenance kits, manual injection valves, mounting kits and syringe.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeFor Use With (Equipment)Description
6042.0600Manual Injection Valve with Mounting Kit2 G UltiMate 3000 PumpBiocompatible, 50 μL PEEK sample loop
6035.0610Mounting Kit1 G UltiMate 3000 Pumpfor Manual Injection Valve
6040.0611Mounting Kit2 G UltiMate 3000 Pumpfor Injection Valve
6040.0110Motorized Injection Valve with Mounting Kit2 G UltiMate 3000 PumpAnalytical/micro
6040.0610Manual Injection Valve with Mounting Kit2 G UltiMate 3000 PumpAnalytical/micro, 20 μL sample loop
Catalog number 6042.0600
Price (INR)
-
Type:
Manual Injection Valve with Mounting Kit
For Use With (Equipment):
2 G UltiMate 3000 Pump
Description:
Biocompatible, 50 μL PEEK sample loop
The Manual Sample Injector Valves and Mounting Kit for Thermo Scientific™ Dionex™ UltiMate™ 3000 Series Pumps include maintenance kits, manual injection valves, mounting kits and syringe.
Specifications
DescriptionBiocompatible, 50 μL PEEK sample loop
For Use With (Equipment)2 G UltiMate 3000 Pump
Biocompatible VersionYes
TypeManual Injection Valve with Mounting Kit
Unit SizeEach