Mixer Kits for UltiMate™ 3000 Pumps
Mixer Kits for UltiMate™ 3000 Pumps
Actual product may vary
Thermo Scientific™
Mixer Kits for UltiMate™ 3000 Pumps
For use with binary, quaternary or dual gradient RS/SD pumps, the Mixer Kits for UltiMate™ 3000 Pumps ensure the highest possible mixing efficiency for ultrahigh resolution and ultrafast separations.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Volume (Metric)
6042.5100Binary, Quaternary, or x2 Dual Gradient RS Pumps100 μL
6040.5000Binary, Quaternary, or x2 Dual Gradient SD Pumps35 μL
6040.5100Binary, Quaternary, or x2 Dual Gradient SD Pumps100 μL
6040.5110Binary, Quaternary, or x2 Dual Gradient SD/RS Pumps200 μL
6040.5310Binary, Quaternary, or x2 Dual Gradient SD/RS Pumps400 μL
6040.5450Binary, Quaternary, or x2 Dual Gradient SD/RS Pumps1550 μL
6040.5750Binary, Quaternary, or x2 Dual Gradient SD/RS Pumps800 μL
6042.5000Binary, Quaternary, or x2 Dual Gradient RS Pumps35 μL
Catalog number 6042.5100
Price (INR)
-
Request A Quote
For Use With (Equipment):
Binary, Quaternary, or x2 Dual Gradient RS Pumps
Volume (Metric):
100 μL
For use with binary, quaternary or dual gradient RS/SD pumps, the Thermo Scientific™ Dionex™ Mixer Kits for Thermo Scientific™ UltiMate™ 3000 Pumps ensure the highest possible mixing efficiency for ultrahigh resolution and ultrafast separations.
Recommended Use
In gradient elution, complete mixing is necessary to achieve a smooth baseline for high signal-to-noise ratios, and accurate peak identification. The mixer requirements are challenging because the mixer must reliably provide highest possible performance over a wide application range with varying solvent ratios, solvent miscibilities, and flow rates. To meet these requirements, the unique SpinFlow™ mixing design was developed, using a two-step process of radial mixing followed by longitudinal mixing. The mixer can be easily adapted to fit specific method requirements, from high-speed LC/MS to high-sensitivity TFA applications.
 Features
  • The two-step process of radial mixing followed by longitudinal mixing ensures the highest possible mixing efficiency.
  • The default mixing volume can be easily changed using mixer kits of 35, 100, 200, 400, 800, or 1550μL (effective mixing volume).
  • The shipment includes all components required for the installation and detailed instructions.
Specifications
DescriptionMixer for 100 μL Mixing Volume
Max. Pressure15000 psi (1034 bar)
Volume (Metric)100 μL
For Use With (Equipment)Binary, Quaternary, or x2 Dual Gradient RS Pumps
Product LineDionex
TypeMixer Kit
Unit SizeEach