Order replacement pistons for your Thermo Scientific™ UltiMate™ 3000 LC Pump when needed.
Have Questions?
Change view
Catalog Number
For Use With (Equipment)
Material
6043.0169
Ultimate 3000 XRS Pumps
Sapphire
6040.0842
Ultimate 3000 BX Pump
Ceramic
2 Options
Catalog number 6043.0169
Price (INR)
-
Request A Quote
For Use With (Equipment):
Ultimate 3000 XRS Pumps
Material:
Sapphire
Order these replacement Thermo Scientific™ Dionex™ UltiMate™ 3000 Pump Pistons when needed. Pistons are available in ceramic and sapphire to meet the needs of your specific pump.