Transcend™ Multichannel Systems
Transcend™ Multichannel Systems
Transcend™ Multichannel Systems
Transcend™ Multichannel Systems
Perform high throughput UHPLC sample preparation, analysis, and clean-up for LC-MS with multichannel UHPLC systems.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
60500-60307Transcend VLX-2 System (F)
60500-60208Transcend LX-4 System
60500-60407Transcend VLX-2 System (H)
Catalog number 60500-60307
Price (INR)
-
Description:
Transcend VLX-2 System (F)
Improve your LC-MS throughput and get faster return on investment with the Thermo Scientific™ Transcend™ Multichannel UHPLC Systems. The unique multichannel technology included within this UHPLC system increases mass spectrometer utilization by engaging up to four separate LC channels combined with one single mass spectrometer. Transcend VLX-2 Systems bring multichannel LC to a new level of simplicity and cost-effectiveness within a single LC stack. Thermo Scientific™ Aria™ Software controls and schedules all aspects of the system; it also features an intuitive graphical user interface for the highest ease of use.
View Transcend System resourcesDiscover VanquishHPLC Support Center
Traditional single channel LC-MS systems process samples sequentially, keeping the MS idle for a significant amount of time. Multichannel LC enables an increased throughput without sacrificing data quality or instrument robustness.
  • Use the cost-effective two channel Transcend VLX-2 System to double your throughput of up to two independent applications without the need for additional bench space while benefiting from the robustness, performance, and ease-of-use built into the system.
  • Use four channel Transcend LX-4 System to increase throughput by up to 400% with optional sample preparation capabilities build into the Triplus autosampler.
Aria Software controls pump operation, valve switching, cleaning, and gradient procedures on the Transcend system. It is compatible with Thermo Scientific™ XCalibur Software or Thermo Scientific™ TraceFinder™ Mass Spectrometer Software, and the intuitive graphical user interface of the Transcend System makes it simple to set up multi-channel methods.
Additional benefits include:
  • Management and control of all aspects of the Transcend system.
  • Scheduling and management of multiple methods on multiple channels, simultaneously.
Specifications
DescriptionTranscend VLX-2 System (F)
For Use With (Application)Double the throughput of any LC-MS analysis or run two LC-MS applications simultaneously.
Max. Flow Rate8 mL/min
For Use With (Equipment)Vanquish Flex Modules
Max. Pressure103 MPa
TypeIntegrated Biocompatible UHPLC System
Unit SizeEach