Order Thermo Scientific™ 6041RS Electrochemical Cell Replacement Gaskets when replacements are needed for your electrochemical cells.
Have Questions?
Change view
Catalog Number
Sample Capacity
6070.2529
50 nL
6070.2528
25 nL
2 Options
Catalog number 6070.2529
Price (INR)
-
Request A Quote
Sample Capacity:
50 nL
Order Thermo Scientific™ 6041RS Electrochemical Cell Replacement Gaskets when replacements are needed for your electrochemical cells. Made of boPET, they are available in 25 and 50nL volumes.
Specifications
DescriptionQuantity: 5/Pk.
For Use With (Equipment)6041 RS Electrochemical Cell