Product Image
Thermo Scientific™
UltiMate™ 3000 Amperometric Electrochemical Cell Electrodes
Find the right electrodes for your amperometric electrochemical analyses utilizing 5040/5041/5041A cells.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeFor Use With (Equipment)
6070.3100Working Electrode, Boron-Doped Diamond6041 RS Cell
70-0096Silver Target Electrode5040 Cell
70-2000Glassy Carbon Target Electrode5040/5041 Cell
70-2134Gold Target Electrode5040 Cell
70-2135Platinum Target Electrode5040 Cell
70-6170Optional Reference Electrode Kit, Ag/AgClModel 5041 Cell
70-6171Optional Reference Electrode Kit, Ag/AgClDual 5041 Cells
6070.3300Working Electrode, Gold6041 RS Cell
6070.3200Working Electrode, Glassy Carbon, High Efficiency6041 RS Cell
70-9030High Sensitivity Glassy Carbon Target Electrode5041 A Cell
Catalog number 6070.3100
Price (INR)
-
Type:
Working Electrode, Boron-Doped Diamond
For Use With (Equipment):
6041 RS Cell
Find the right Thermo Scientific™ Dionex™ UltiMate™ 3000 Amperometric Electrochemical Cell Electrodes for your amperometric electrochemical analyses utilizing 5040/5041/5041A cells. They are available as target, reference or working electrodes for a variety of electrochemical analyses.
Specifications
For Use With (Equipment)6041 RS Cell
TypeWorking Electrode, Boron-Doped Diamond
Unit SizeEach