UltiMate™ 3000 Diode Array Detector Flow Cells
Actual product may vary
UltiMate™ 3000 Diode Array Detector Flow Cells
UltiMate™ 3000 Diode Array Detector Flow Cells
UltiMate™ 3000 Diode Array Detector Flow Cells
UltiMate™ 3000 Diode Array Detector Flow Cells
Thermo Scientific™
UltiMate™ 3000 Diode Array Detector Flow Cells
Choose the flow cell with the desired flow rate for the Thermo Scientific™ Dionex™ UltiMate™ 3000 Diode Array Detector.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeMaterialVolume (Metric)
6082.0400Analytical Flow CellPEEK13 μL
6082.0300Semi-Micro Flow CellStainless Steel2.5 μL
6082.0500Semi-Micro Flow CellPEEK2.5 μL
6082.0100Analytical Flow CellStainless Steel13 μL
6082.0200Semi-Micro Flow CellStainless Steel5 μL
6082.0600Semi-Prep Flow CellPEEK2.5 μL
Catalog number 6082.0400
Price (INR)
-
Type:
Analytical Flow Cell
Material:
PEEK
Volume (Metric):
13 μL
Choose the Thermo Scientific™ UltiMate™ 3000 Diode Array Detector Flow Cell with the desired flow rate. Flow cells are available in analytical, semi-prep and semi-micro flow rates.
Flow cells are available in both stainless steel and PEEK to meet your application requirements.
Specifications
MaterialPEEK
Path Length10 mm light path
Volume (Metric)13 μL
For Use With (Equipment)Dionex UltiMate 3000 Diode Array Detector
Product LineUltiMate
TypeAnalytical Flow Cell
Unit SizeEach