Vanquish™ LightPipe™ Flow Cells for the Vanquish™ Diode Array Detector HL
Vanquish™ LightPipe™ Flow Cells for the Vanquish™ Diode Array Detector HL
Actual product may vary
Thermo Scientific™
Vanquish™ LightPipe™ Flow Cells for the Vanquish™ Diode Array Detector HL
Obtain outstanding S/N performance and reduced interfering RI and thermal effects using these flow cells for the Thermo Scientific™ Vanquish™ Diode Array Detector.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionType
6083.0300LightPipe diagnostic cellDiagnostic Cell
6083.0100BLightPipe flow cell, 10 mmStandard
6083.0200BLightPipe flow cell, 60 mm6167028
Catalog number 6083.0300
Price (INR)
-
Request A Quote
Description:
LightPipe diagnostic cell
Type:
Diagnostic Cell
The completely new design of the Thermo Scientific™ Vanquish™ Diode Array Detector (DAD) HL incorporates Thermo Scientific™ LightPipe™ Flow Cells, which provide the best signal-to-noise (S/N) performance through the combination of lowest baseline noise, a very long light path, and minimum peak dispersion. The ultra-wide dynamic range is ideal for simultaneous detection of highly concentrated main compounds and impurities down to trace levels. Interfering refractive index (RI) and thermal effects are reduced to a minimum for less baseline drift and therefore more reliable peak integration.
Great standards in flow cells for a wide range of applications
Standard flow cell:
  • Volumes: 2μL (illuminated), 0.8μL (dispersion)
  • Flow Cell Pressure Limit 6MPa (60bar, 870psi)
High sensitivity flow cell:
  • Volumes: 13μL (illuminated), 4.0μL (dispersion)
  • Flow Cell Pressure Limit 6MPa (60bar, 870psi)
Specifications
DescriptionLightPipe diagnostic cell
For Use With (Equipment)Vanquish Diode Array Detector HL
Product LineLightPipe
TypeDiagnostic Cell
Unit SizeEach