Product Image
Bath Fluids for Thermo Scientific Neslab Baths
Use ethylene glycol for low-temperature applications to -30°C, silicone oil for high temperature applications to +150°C.
Have Questions?
Catalog NumberType
610000000001Ethylene Glycol Bath Fluid
Catalog number 610000000001
Price (INR)
-
Type:
Ethylene Glycol Bath Fluid
Use ethylene glycol for low-temperature applications to -30°C, silicone oil for high temperature applications to +150°C.
Use ethylene glycol for low-temperature applications to -30°C, silicone oil for high temperature applications to +150°C.
Specifications
For Use With (Application)Neslab bath
Capacity (Metric)19 L
Temperature Range-30°C to +150°C
TypeEthylene Glycol Bath Fluid
Unit SizeEach