Product Image
Thermo Scientific™
Pass-Thru Port for Ultralow and Cryogenic Freezers
Pass probes, cables, leads or measuring instruments through the Thermo Scientific™ Pass-Thru Port for Ultralow and Cryogenic Freezers.
Have Questions?
Catalog number 6105-1
Price (INR)
-
Pass probes, cables, leads or measuring instruments through the Thermo Scientific™ Pass-Thru Port for Ultralow and Cryogenic Freezers.
  • 1 in. (2.5cm) dia. pass-through ports are standard on ultra low-temperature and cryogenic freezers
  • Additiional ports are available on some models
Specifications
DescriptionPass-Thru port; 1in. (2.5cm) dia.
For Use With (Equipment)Ultra-Low Temperature and Cryogenic Freezers
TypePass-Thru Port
Unit SizeEach