Fluid Filter Kit
Actual product may vary
Thermo Scientific™
Fluid Filter Kit
Fluid Filter Kits are available with 5 to 40μm partial-flow filtration.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeFor Use With (Equipment)Partial Flow
611000000096Full Flow Fluid FilterMerlin M100 thru M15010 micron
611000000084Partial Flow Fluid Filter KitMerlin M25 thru M1505 micron
611000000085Partial Flow Fluid FilterMerlin M25 thru M15010 micron
611000000086Partial Flow Fluid FilterMerlin M25 thru M15025 micron
611000000087Partial Flow Fluid FilterMerlin M25 thru M15040 micron
611000000097Full Flow Fluid FilterMerlin M100 thru M15025 micron
611000000098Full Flow Fluid FilterMerlin M100 thru M15040 micron
611000000095Full Flow Fluid FilterMerlin M100 thru M1505 micron
280010051100Partial Flow Fluid FilterMerlin M25 thru M1505 micron
627000000010Fluid StrainerMerlin Water Baths100 mesh
611000000101Replacement Filter BagsThermoFlex 7500 or 2400035 micron
611000000102Replacement Filter BagsThermoFlex 7500 or 24000100 micron
598000000015Replacement Filter BagsThermoFlex 7500 or 24000250 micron
Catalog number 611000000096
Price (INR)
-
Type:
Full Flow Fluid Filter
For Use With (Equipment):
Merlin M100 thru M150
Partial Flow:
10 micron
Thermo Scientific™ Fluid Filter Kits are available with 5 to 40μm partial-flow filtration.
Specifications
For Use With (Equipment)Merlin M100 thru M150
Partial Flow10 micron
Size3/4 in.
TypeFull Flow Fluid Filter
Unit SizeEach