Sterilin™ Clear Microtiter™ Plates
Actual product may vary
Thermo Scientific™
Sterilin™ Clear Microtiter™ Plates
Store and transport samples or perform microbiology screenings, EIA/absorbance assays and serology work with 96-well Thermo Scientific™ Sterilin™ Clear Microtiter™ Plates.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeWell ShapeSterility
611V96Microtiter PlatesConical Bottom (V-well)Non-sterile
611U96Microtiter PlatesRound Bottom (U-well)Non-sterile
611F96Microtiter PlatesFlat Bottom (F-well)Non-sterile
612U96Microtiter PlatesRound Bottom (U-well)Sterile
612V96Microtiter PlatesConical Bottom (V-well)Sterile
612F96Microtiter PlatesFlat Bottom (F-well)Sterile
642000Plate LidNot ApplicableSterile
Catalog number 611V96
Price (INR)
-
Product Type:
Microtiter Plates
Well Shape:
Conical Bottom (V-well)
Sterility:
Non-sterile
Store and transport samples or perform microbiology screenings, EIA/absorbance assays and serology work with 96-well Thermo Scientific™ Sterilin™ Clear Microtiter™ Plates. Supplied without lid; may be purchased separately. Not available in all geographic regions.
Features
  • Manufactured from virgin polystyrene for excellent optical clarity
  • Manufactured in ISO Class 7 Clean Room without surface treatment for a medium binding surface
  • Available irradiated for sterility and a more hydrophilic surface, providing high binding
  • Suitable for use at a wavelength of 340nm
  • Minimizes the risk of contamination from surrounding wells with condensation rings on the lid raised well rims on the base.
  • Ensures even temperature distribution around each well with a hanging well design
Easy Sample Identification
  • Frosted writing area on the end wall
  • Orientation corners
  • Alphanumeric labeling
Note:
Plates are supplied without a lid (can be purchased separately).
Ordering Information:
Product is not available in all geographic regions.
Specifications
Certifications/ComplianceCE marked in accordance with the European Directive 98/72/EC
FormatStandard
No. per Case50
No. per Pack5
Packaging50 Cases
Product TypeMicrotiter Plates
Quantity50/Pk.
Array8 x 12
ColorClear
MaterialPolystyrene
No. of Wells96
SterilityNon-sterile
Surface TreatmentNon-treated
Well ShapeConical Bottom (V-well)
Unit SizeEach