Drawer Dividers
Actual product may vary
Thermo Scientific™
Drawer Dividers
Maximize storage inside blood bank refrigerators and plasma freezers with these optional drawer dividers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
620623.3, 51.1 and 78.8 cu. ft. Thermo Scientific Revco and Forma blood bank refrigerators and plasma freezers.
696329.2 cu. ft. Thermo Scientific Revco and Forma blood bank refrigerators and plasma freezers.
Catalog number 6206
Price (INR)
-
For Use With (Equipment):
23.3, 51.1 and 78.8 cu. ft. Thermo Scientific Revco and Forma blood bank refrigerators and plasma freezers.
Maximize storage inside blood bank refrigerators and plasma freezers with these optional Thermo Scientific™ Drawer Dividers.
Stainless-steel drawer dividers help manage inventory of bags and boxes:
  • Four column dividers for all models except 29.2 cu. ft. (826L) models
  • Five column dividers for 29.2 cu. ft. (826L) models
  • Warranty varies by country, please contact us for details
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description4-column divider
DrawersStainless Steel (Drawer)
For Use With (Equipment)23.3, 51.1 and 78.8 cu. ft. Thermo Scientific Revco and Forma blood bank refrigerators and plasma freezers.
HoldsBoxes
Type4-Column Divider
Capacity (English)78.8 cu. ft.
Capacity (Metric)2231.3 L
Unit SizeEach