Search
| Catalog Number | Width (English) |
|---|---|
| 6283-1850PK also known as 6283-1850 | 18 in. |
Find a variety of uses for Thermo Scientific™ Nalgene™ CleanSheets™ Polyethylene Bench/Drawer Liners, such as sink liners or titration backgrounds. These liners can be cut to any size and stapled, tacked or taped. They provide cushioning and liquid barrier protection.
Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in.