You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture and Transfection
Chemicals
Chromatography
Electron Microscopes
Lab Plasticware and Supplies
Lab Centrifuges
Lab Solutions
Mass Spectrometers
Next Generation Sequencers
See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
New Products
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
Home
▸
Shop All Products
▸
Tubes
▸
PCR Tubes
▸
HotStart™ Storage Reaction Tubes
Actual product may vary
Thermo Scientific™
HotStart™ Storage Reaction Tubes
Eliminate tedious oil or wax overlays with the thin-walled Thermo Scientific™ HotStart™ Storage Reaction Tubes.
Have Questions?
Change view
Catalog Number
Product Line
Volume (Metric)
6310-SDG
Hot Start Micro 50 Bulk
0.2 mL
6702TS
also known as
6702
Hot Start Micro 50 Bulk
0.5 mL
2 Options
Catalog number
6310-SDG
Price (INR)
-
Request A Quote
Product Line:
Hot Start Micro 50 Bulk
Volume (Metric):
0.2 mL
Eliminate tedious oil or wax overlays with the thin-walled Thermo Scientific™ HotStart™ Storage Reaction Tubes.
Features:
Bead is stable at room temperature
Assures synchronous reaction startup
PCR product can remain in tube for later analysis
Hinged caps provide snug seal
Presterilized; RNase- and DNase-free
Recommended for:
Complex DNA or cDNA, very low copy number targets, more than 30 thermal cycles, multiple PCR or RT-PCR
Specifications
For Use With (Equipment)
Thermal Cyclers
Product Line
Hot Start Micro 50 Bulk
Reaction Volume
25 to 50 μL
Product Type
Tube
Sterility
Sterile
Volume (Metric)
0.2 mL
Unit Size
Each