Cryoware Permanent Markers and Pens
Cryoware Permanent Markers and Pens
Actual product may vary
Cryoware Permanent Markers and Pens
Cryoware Permanent Markers and Pens
Cryoware Permanent Markers and Pens

Cryoware Permanent Markers and Pens

Identify samples and storage boxes permanently with these cryoware permanent markers and pens.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorNo. per Case
6313-0020Black4
339993Assorted (Red, Blue, Green, Black)100
6313-0010Assorted (Red, Blue, Green, Black)24
Catalog number 6313-0020
HSN 96082000
Price (INR)
26,626.00
Case
Add to cart
Color:
Black
No. per Case:
4
Price (INR)
26,626.00
Case
Add to cart

Permanently notate samples and storage boxes with Thermo Scientific™ Cryoware Permanent Markers and Pens, which can easily withstand cold storage temperatures.

 Warranty: 90 days
Specifications
Product TypePermanent Marking Pen
Tip StyleExtra Fine
ColorBlack
DescriptionCryoware Permanent Marker
IncludesConsists of four extra-fine-point pens
For Use With (Application)Used for marking cardboard and Polycarbonate
No. per Case4
Quantity1 Pk.
Unit SizeCase