Search
| Catalog Number | Material | Closure Size |
|---|---|---|
| 6421-0010 | Polypropylene | 1.12 in. x 12 female screw thread |
| 6420-0750 | LDPE | 19mm NPT male screw thread |
Ensure safe and efficient liquid dispensing from tanks by using Thermo Scientific™ Nalgene™ Spigots for Storage Tanks. Spigots are for use with Naglene™ tanks up to 100 gal. with factory installed threaded boss.
Includes
Warranty:
90 days
Note:
Make sure container and spigot have compatible chemical resistance before installing.
Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.