VersaTREK™ Starter Kit
Thermo Scientific™
VersaTREK™ Starter Kit
Facilitate efficiency and get up and running with ease with a complete kit of accessories for the Thermo Scientific™ VersaTREK™ Automated Microbial Detection System.
Have Questions?
Catalog NumberPackaging
6499-31Ea.
Catalog number 6499-31
Price (INR)
-
Packaging:
Ea.
Facilitate efficient operation of the Thermo Scientific™ VersaTREK™ System right at the start with a full line of instrument accessories packed into a single convenient kit. The Thermo Scientific VersaTREK Starter Kit includes Disabled Location Caps (5), Stylus (pack of 4), Bottle Carrying Rack (1), Thermometer (3), Thermometer Mounts (6), Absorbent Sheets (3), and Labels for Drawer Numbers 1-22 (1).
Meet the demands of both high and low volume laboratories with the Thermo Scientific VersaTREK System, which comes with a full line of ancillary and accessory products. With four FDA-cleared tests and a simple, two-bottle media system, the VersaTREK System provides maximum value with faster results and fewer limitations.
  • Only system capable of detecting any gas produced or consumed by organisms, allowing detection of a wider range of both common and fastidious organisms
  • Simplified workflow includes easy scan function for bottle entry, intuitive icons to reduce training time, and no daily QC requirement for maximum efficiency
  • Highly-enriched Thermo Scientific VersaTREK REDOX™ media supports a wide range of organisms, facilitating better recovery of fastidious organisms
Kit includes: (5) 6044-30, (1) 6420-30, (1) 6424-30, (3) 6440-30, (3) 6444-30, (1) 6452-30 & (6) 6705-30.
Not all products are available for sale in all territories. Please inquire.
VersaTREK, Sensititre™, Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products with a comprehensive line of media and diagnostic products, providing a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
DescriptionVersaTREK Starter Kit; Includes (5) 6044-30, (1) 6420-30, (1) 6424-30, (3) 6440-30, (3) 6444-30, (1) 6452-30 & (6) 6705-30
PackagingEa.
Product TypeVersaTREK Starter Kit
Unit SizeEach