Power Supply Unit (24V) for UltiMate™ 3000 Solvent Racks
Power Supply Unit (24V) for UltiMate™ 3000 Solvent Racks
Thermo Scientific™

Power Supply Unit (24V) for UltiMate™ 3000 Solvent Racks

Solvent Rack Power Supply for UltiMate 3000 Solvent Racks is designed for use with SOR-100 and SRD-3x00 pumps.
Have Questions?
Catalog NumberFor Use With (Equipment)
6510.0004UltiMate 3000 Solvent Racks
Catalog number 6510.0004
Price (INR)
-
Request A Quote
For Use With (Equipment):
UltiMate 3000 Solvent Racks

Choose Thermo Scientific™ Solvent Rack Power Supply for UltiMate™ 3000 Solvent Racks designed for use with SOR-100 and SRD-3x00 pumps.

24V
Specifications
DescriptionPower Supply Unit 24 V for SOR-100 and SRD-3x00
For Use With (Equipment)UltiMate 3000 Solvent Racks
TypePower Supply Unit
Unit SizeEach