Dynabeads™ Streptavidin Magnetic Beads
Invitrogen™

Dynabeads™ Streptavidin Magnetic Beads

Dynabeads Streptavidin Magnetic Beads are designed for high-specificity, low-background capture of target molecules with biotinylated affinity binders such as antibodies, antigens, or oligonucleotides. Available in various sizes and surface chemistries, these beads can be tailored to provide the best fit for specific application needs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeQuantity
65607DDynabeads™ Streptavidin for Target Enrichment50 mL
65001Dynabeads™ MyOne™ Streptavidin C12 mL
65002Dynabeads™ MyOne™ Streptavidin C110 mL
65604DDynabeads™ MyOne™ Streptavidin T150 mL
65602Dynabeads™ MyOne™ Streptavidin T110 mL
65601Dynabeads™ MyOne™ Streptavidin T12 mL
65605DDynabeads™ Streptavidin for Target Enrichment2 mL
65606DDynabeads™ Streptavidin for Target Enrichment10 mL
11205DDynabeads™ M-280 Streptavidin2 mL
11206DDynabeads™ M-280 Streptavidin10 mL
60210Dynabeads™ M-280 Streptavidin100 mL
65305Dynabeads™ M-270 Streptavidin2 mL
65306Dynabeads™ M-270 Streptavidin10 mL
60101Dynabeads™ kilobaseBINDER™ Kit200 Isolations
65801DDynabeads™ Streptavidin Trial Kit4 x 1 mL
Catalog number 65607D
Price (INR)
556,134.00
Online offer
Ends: 31-Dec-2026
823,902.00
Save 267,768.00 (33%)
Each
Product Type:
Dynabeads™ Streptavidin for Target Enrichment
Quantity:
50 mL
Price (INR)
556,134.00
Online offer
Ends: 31-Dec-2026
823,902.00
Save 267,768.00 (33%)
Each

Dynabeads Streptavidin Magnetic Beads are designed for high-specificity, low-background capture of target molecules with biotinylated affinity binders such as antibodies, antigens, or oligonucleotides. Available in various sizes and surface chemistries, these beads can be tailored to provide the best fit for specific application needs.

Dynabeads Streptavidin Magnetic Beads are compatible with high-throughput automation platforms, including KingFisher sample purification systems, ensuring seamless integration into new and existing workflows.

Key advantages of Dynabeads Streptavidin Magnetic Beads

  • ISO 13485–certified manufacturing—exceptional manufacturing control to ensure robust and consistent performance between production lots, meeting international quality standards for medical devices
  • Monosized, monodispersed magnetic beads—uniform particle size provides consistent and reproducible results across applications
  • Dual size options—available in 1-μm and 2.8-μm diameter specifications to accommodate diverse experimental requirements and application-specific needs
  • Multiple surface chemistry options—various surface functionalization chemistries available to optimize binding efficiency and compatibility for specific applications
  • Application-optimized design—size and surface chemistry combinations enable best-fit selection for targeted experimental protocols and assay requirements

Precision-engineered manufacturing delivers exceptional control over critical bead properties

Dynabeads Streptavidin products are based on magnetic polystyrene-based beads that are covalently coupled with recombinant streptavidin, creating a robust platform for target capture and isolation using biotinylated affinity binders. Dynabeads Streptavidin product manufacturing enables exceptional precision over three critical parameters: size distribution, magnetization properties, and surface chemical characteristics. This ensures consistent performance across batches and applications, providing the reliability necessary for reproducible experimental outcomes.

Optimized size selection enhances application-specific performance

The Dynabeads Streptavidin product line offers two distinct size options, each engineered to excel in specific experimental contexts. The 1-μm beads, available in the Dynabeads MyOne series, are ideally suited for applications requiring fast reaction kinetics, slow sedimentation rates, and high sensitivity-to-capacity ratios. These smaller beads provide enhanced surface area contact and reduced gravitational settling, making them particularly valuable for sensitive detection assays.

In contrast, the 2.8-μm beads, found in the Dynabeads M-270 and M-280 series, are perfect for applications demanding low background binding and reduced risk of aggregation in aggregation-prone applications. These larger beads offer even easier handling and faster magnetic separation compared to the Dynabeads MyOne series, facilitating streamlined workflow processing.

Tailored surface chemistry options address diverse experimental requirements

The Dynabeads Streptavidin product line includes two distinct surface chemistry configurations designed to optimize performance across varied experimental conditions. Hydrophobic surfaces are available in Dynabeads MyOne Streptavidin T1, Dynabeads Streptavidin for Target Enrichment, and Dynabeads M-280 Streptavidin products. These surfaces are BSA-blocked after streptavidin coupling making them ideal for high signal-to-noise applications such as immunoassays, target enrichment, and on-bead PCR for next-generation sequencing.

Alternatively, hydrophilic surfaces are featured in Dynabeads MyOne Streptavidin C1, Dynabeads Streptavidin for In Vitro Transcription, and Dynabeads M-270 Streptavidin products. These surfaces contain no BSA blocking and are particularly suitable for hydrophobic targets, harsh lysis conditions, sample preparation workflows for mass spectrometry, and applications demanding low or no nuclease activity, such as on-bead reverse transcription or in vitro transcription.

For researchers uncertain about which bead type best suits their application, a Dynabeads Streptavidin Trial Kit is available. It allows for side-by-side performance evaluation of different bead formulations under your specific experimental conditions, ensuring optimal selection for your research needs.

ISO 13485 certification ensures scalable quality and OEM support

Manufactured under ISO 13485 certification, Dynabeads Streptavidin products deliver outstanding batch-to-batch consistency. This robust quality assurance framework ensures that once customers develop an assay or workflow, they can rely on reproducible results across multiple experimental iterations. This consistency makes these versatile beads an efficient solution for applications spanning the entire spectrum from fundamental research to commercial manufacturing processes. For researchers developing commercial products or services, or those requiring larger bulk volumes, specialized support is available by emailing oemdynal@thermofisher.com.

For Research Use Only.
Specifications
Bead TypeMagnetic polystyrene-based beads covalently coupled with recombinant Streptavidin
Binding Property950-1500 pmoles free biotin/mg beads
Certifications/ComplianceISO9001 and ISO13485
Concentration10 mg/mL
Diameter (Metric)1 μm (CV < 5%)
For Use With (Application)Capture of targeted DNA sequences for Next-Generation Sequencing (NGS) and hybridization-based DNA assays
For Use With (Equipment)KingFisher™ Sample Purification System, DynaMag™ magnets
High-throughput CompatibilityHigh-throughput Compatible
Product LineDynabeads
Product TypeDynabeads™ Streptavidin for Target Enrichment
Quantity50 mL
Regulatory StatusRUO
Shelf Life48 months from date of manufacture
Shipping ConditionAmbient Temperature
Surface FunctionalityTosylactivated hydrophobic surface blocked with BSA
FormatBeads in Suspension
Isolation TechnologyMagnetic Bead
Unit SizeEach
Contents & Storage
Dynabeads Streptavidin for Target Enrichment is supplied in PBS, pH 7.4, with 0.1% BSA, 0.02% NaN3. Store at 2°C to 8°C.

Frequently asked questions (FAQs)

What range of DNA fragment sizes can be captured with Dynabeads Streptavidin for Target Enrichment (Cat. No. 65605D)?

We do not have hard cutoffs for the lengths of the fragments that can be captured with our Dynabeads products, including Cat. No. 65605D. In general terms, however, the beads should work well with any fragment up to 2kb. If you have mixed sizes in the sample, you can expect the smaller fragments being captured more efficiently than the longer ones.

Find additional tips, troubleshooting help, and resources within our Dynabeads Nucleic Acid Purification Support Center.