Slide-A-Lyzer™ Dialysis Cassettes, 10K MWCO
Thermo Scientific™

Slide-A-Lyzer™ Dialysis Cassettes, 10K MWCO

Remove salts and small contaminants to maximize sample purity and recovery with Slide-A-Lyzer 10K MWCO dialysis cassettes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSample Volume (Metric)QuantityProduct Type
663803 mL10 CassettesDialysis Cassettes
663830.5 mL10 CassettesDialysis Cassettes
663840.5 mL5 x 10 CassettesDialysis Cassettes
663850.5 mL10 CassettesDialysis Cassette Kit
663813 mL5 x 10 CassettesDialysis Cassettes
663823 mL10 CassettesDialysis Cassette Kit
6681012 mL8 CassettesDialysis Cassettes
6681112 mL5 x 8 CassettesDialysis Cassettes
6680712 mL8 CassettesDialysis Cassette Kit
6683030 mL6 CassettesDialysis Cassettes
664540.5 mL10 CassettesDialysis Cassettes (gamma-irradiated)
664553 mL10 CassettesDialysis Cassettes (gamma-irradiated)
6645312 mL8 CassettesDialysis Cassettes (gamma-irradiated)
6645630 mL6 CassettesDialysis Cassettes (gamma-irradiated)
Catalog number 66380
Price (INR)
23,177.00
Each
Sample Volume (Metric):
3 mL
Quantity:
10 Cassettes
Product Type:
Dialysis Cassettes
Request bulk or custom format
Price (INR)
23,177.00
Each
Slide-A-Lyzer Dialysis Cassettes (10K MWCO) facilitate the simple and effective removal of salts and small contaminants from proteins and other macromolecules greater than 10,000 Daltons. Slide-A-Lyzer Dialysis Cassettes are available in four sizes to accommodate sample volumes between 0.5 and 30 mL. These devices are easier to use than traditional dialysis tubing, providing faster buffer exchange and more complete sample recovery.
Slide-A-Lyzer cassettes can be used for a wide range of applications including low-molecular weight contaminant removal, buffer exchange, desalting, equilibrium dialysis, and sample concentration. Liquid samples are easily added and removed by penetrating the self-sealing gasket with a hypodermic needle attached to a syringe. No knots, clips, or caps are needed to seal the units and prevent leaking and sample-loss during the dialysis procedure.

Slide-A-Lyzer Dialysis Cassettes are constructed from two sheets of low-binding, regenerated-cellulose dialysis membrane that are hermetically sealed on either side of a silicone-like gasket inside an inert plastic frame. The membrane and cassette materials are compatible with most common laboratory chemicals and buffers.

Features of Slide-A-Lyzer Dialysis Cassettes:
Color-coded—orange frame specifies cassettes with the 10,000 MW pore size membrane
Four cassette sizes—select the cassette that best suits your sample volume
Fast dialysis—flat cassette chamber with two membranes provides high surface area-to-volume ratio that maximizes diffusion rate compared to cylindrical dialysis tubing
High recovery—rectangular cassette design maximizes recovery of entire sample volume via any one of the four corner injection ports
Gamma-irradiated option—gamma-irradiated cassettes are available for applications requiring conditions that minimize the risk of sample contamination

Gamma-irradiated dialysis cassettes
Gamma-irradiated Slide-A-Lyzer Dialysis Cassettes are ideal for researchers culturing cells and microorganisms; purifying viruses, DNA, and RNA; or performing sample preparation for other applications requiring conditions that minimize the risk of sample contamination. The gamma-irradiated cassettes are treated with 20–36 kGy of gamma radiation, a dosage that has been determined to sterilize the units based on both trypticase soy broth medium for identifying total growth and fluid thioglycollate medium for identifying anaerobic growth. However, a confirmation of sterility is not provided for individual lots, as no further sterility testing is conducted post-irradiation.

Membrane specifications:
Composition: regenerated cellulose synthesized by the viscose method
Thickness: 0.9 to 1.2 mil (22.5 to 30 μm)
Hydration time required before use: 2 minutes
Glycerol content: 21%
Sulfur content: 0.1–0.15%
Heavy metals content: trace

Volume ranges:
• 0.5 mL = 0.1 to 0.5 mL
• 3 mL = 0.5 to 3 mL
• 12 mL = 3 to 12 mL
• 30 mL = 12 to 30 mL

For Research Use Only. Not for use in diagnostic procedures.
Specifications
MembraneCellulose Membrane, Dialysis
Product LineSlide-A-Lyzer
Product TypeDialysis Cassettes
Purification TargetBuffer Exchange, Protein
Quantity10 Cassettes
FormatCassette
MWCO10 kDa
Sample Volume (Metric)3 mL
Unit SizeEach
Contents & Storage
Store at room temperature

Frequently asked questions (FAQs)

Does the Slide-A-Lyzer Dialysis Cassette leak?

The innovative, patented design does not permit “wicking” that can occur with homemade microdialysis devices.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

What sizes are available for the Slide-A-Lyzer Dialysis Cassette? What MWCO membranes are available?

Four sizes to accommodate samples between 0.1 and 30 ml are currently available in five different MWCOs (2K, 3.5K, 7K, 10K and 20K). The G2 Slide A lyzers on the other hand are available in five different sizes to accommodate samples between 0.25 and 70mL.
These units, especially those that accommodate smaller volumes, provide for greater sample recovery and shorter dialysis times than one can achieve with traditional tubing. We have a cassette selection guide to help you choose the appropriate cassette for your application. There are also product demonstrations for this and other High-Performance Dialysis Products.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

What prevents the Slide-A-Lyzer Dialysis Cassette from leaking?

An internal gasket, which isn't visible after the cassette is assembled, reseals itself as the needle is withdrawn. To maintain sample volume and integrity, users should remember these three key rules:

1. Before use, fill the cassette with air to verify that the system is airtight.
2. Puncture each porthole only once.
3. Do not allow the membrane to come into contact with the sharp needle bevel.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

What is the purpose of the buoy for the Slide-A-Lyzer Dialysis Cassette?

The buoy suspends the Slide-A-Lyzer Cassette above the stirring bar during dialysis and it provides a benchtop stand on which to set the Slide-A-Lyzer Cassette. These are not required with the G2 Slide-A-Lyzer Dialysis Cassettes.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

What is the highest temperature and the pH range that is compatible with the Slide-A-Lyzer Cassette?

Slide-A-Lyzer Cassettes can be held at 56 degrees C for two hours. At temperatures and times above these, the plastic gets soft and can deform. Thermo Scientific recommends that users maintain a pH range of 5-9.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Citations & References (8)

Citations & References
Abstract
Tyrosine-Based Cross-Linking of Peptide Antigens to Generate Nanoclusters with Enhanced Immunogenicity: Demonstration Using the Conserved M2e Peptide of Influenza A.
Authors:Wilks LR,Joshi G,Grisham MR,Gill HS
Journal:ACS infectious diseases
PubMed ID:34432416
A method of creating nanoclusters (NCs) from soluble peptide molecules is described utilizing an approach based on a tyrosine-tyrosine cross-linking reaction. A reactive tag comprising histidine and tyrosine residues was introduced at the termini of the peptide molecules. The cross-linking reaction led to the creation of dityrosine bonds within the ... More
descSPIM: an affordable and easy-to-build light-sheet microscope optimized for tissue clearing techniques.
Authors:Otomo K,Omura T,Nozawa Y,Edwards SJ,Sato Y,Saito Y,Yagishita S,Uchida H,Watakabe Y,Naitou K,Yanai R,Sahara N,Takagi S,Katayama R,Iwata Y,Shiokawa T,Hayakawa Y,Otsuka K,Watanabe-Takano H,Haneda Y,Fukuhara S,Fujiwara M,Nii T,Meno C,Takeshita N,Yashiro K,Rosales Rocabado JM,Kaku M,Yamada T,Oishi Y,Koike H,Cheng Y,Sekine K,Koga JI,Sugiyama K,Kimura K,Karube F,Kim H,Manabe I,Nemoto T,Tainaka K,Hamada A,Brismar H,Susaki EA
Journal:Nature communications
PubMed ID:38866781
Despite widespread adoption of tissue clearing techniques in recent years, poor access to suitable light-sheet fluorescence microscopes remains a major obstacle for biomedical end-users. Here, we present descSPIM (desktop-equipped SPIM for cleared specimens), a low-cost ($20,000-50,000), low-expertise (one-day installation by a non-expert), yet practical do-it-yourself light-sheet microscope as a solution ... More
Determination of seroprevalence and kinetics of humoral response using mpox virus A29 protein.
Authors:Cai JP,Chu WM,Tam AR,Wang K,Han Y,Chen LL,Zhang X,Choi CY,Cheng VC,Chan KH,Chen Z,Hung IF,Fong CH,To KK
Journal:Communications medicine
PubMed ID:37993690
BACKGROUND: Mpox virus (MPXV), previously known as monkeypox virus, has spread globally in 2022. An accurate and convenient antibody test is essential for the determination of seroprevalence and for studying immune response after natural infection or vaccination. Most seroprevalence or vaccine studies used either live MPXV (or vaccinia virus [VACV]) or ... More
ADIBO-based "click" chemistry for diagnostic peptide micro-array fabrication: physicochemical and assay characteristics.
Authors:Prim D,Rebeaud F,Cosandey V,Marti R,Passeraub P,Pfeifer ME
Journal:Molecules (Basel, Switzerland)
PubMed ID:23959194
Several azide-derivatized and fluorescently-labeled peptides were immobilized on azadibenzocyclooctyne (ADIBO)-activated slide surfaces via a strain-promoted alkyne-azide cycloaddition (SPAAC) reaction revealing excellent immobilization kinetics, good spot homogeneities and reproducible fluorescence signal intensities. A myc-peptide micro-array immunoassay showed an antibody limit-of-detection (LOD) superior to a microtiter plate-based ELISA. Bovine serum albumin (BSA) ... More
Development of a high-throughput enzyme-linked immunosorbent assay for the routine detection of the carcinogen acrylamide in food, via rapid derivatisation pre-analysis.
Authors:Preston A,Fodey T,Elliott C
Journal:Analytica chimica acta
PubMed ID:18215649
The spontaneous formation of the neurotoxic carcinogen acrylamide in a wide range of cooked foods has recently been discovered. These foods include bread and other bakery products, crisps, chips, breakfast cereals, and coffee. To date, the diminutive size of acrylamide (71.08 Da) has prevented the development of screening immunoassays for ... More