Search
| Catalog Number | For Use With (Equipment) | Type |
|---|---|---|
| 6695-1 | 29.2 cu. ft. High-performance Units | Open Wire Shelves |
| 6695 | 29.2 cu. ft. Units | Open Wire Shelves |
| 6696 | 23.3, 45.8, 51.1, 78.8 cu. ft. Units | Open Wire Shelves |
| 6696-1 | 23.3, 45.8, 51.1 cu.ft. Units | Full Solid Shelves |
| 6697 | 29.2 cu. ft. Units | Open Wire Shelves |
| 6698 | 51.1 cu. ft. Units | Open Wire Shelves |
Maximize lab storage with Thermo Scientific™ Freezer and Refrigerator Shelves, designed for use as replacement shelves in high-performance laboratory refrigerators, freezers and ULTs.