Resurge™ CD2 Supplement, without glucose and L-glutamine
Actual product may vary
Gibco™
Resurge™ CD2 Supplement, without glucose and L-glutamine
Resurge CD2 Supplement, without glucose and L-glutamine, is a chemically defined supplement formulation to complement a variety of base media.Read more
Have Questions?
Change view
Catalog Number
Quantity
670017
5 kg
670015
100 g
670016
1 kg
3 Options
Catalog number 670017
Price (INR)
-
Request A Quote
Quantity:
5 kg
Resurge CD2 Supplement, without glucose and L-glutamine, is a chemically defined supplement formulation to complement a variety of base media. This supplement was developed using several host cell lines and in various culture systems, including shaker flasks and bench-scale bioreactors. Resurge CD2 Supplement, without glucose and L-glutamine, was developed for use in CHO mammalian culture systems and can be used either as a media supplement or a culture system feed.