Resurge™ CD4 Supplement, without glucose and L-glutamine
Actual product may vary
Gibco™
Resurge™ CD4 Supplement, without glucose and L-glutamine
Resurge CD4 Supplement, without glucose and L-glutamine, is a chemically defined supplement formulation to complement a variety of base media.Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
6700221 kg
670021100 g
Catalog number 670022
Price (INR)
-
Quantity:
1 kg
Resurge CD4 Supplement, without glucose and L-glutamine, is a chemically defined supplement formulation to complement a variety of base media. This supplement was developed using several host cell lines and in various culture systems, including shaker flasks and bench-scale bioreactors. Resurge CD4 Supplement, without glucose and L-glutamine, was developed for use in CHO mammalian culture systems and can be used either as a media supplement or a culture system feed.
Specifications
Cell LineCHO
Culture TypeMammalian Cell Culture
FormatBottle(s)
Product LineResurge
Quantity1 kg
SourceAnimal origin-free
ClassificationAnimal Origin-free, Chemically-defined, Protein-free
FormPowder
Product TypeMedia Supplement
Without AdditivesNo Glutamine, No Glucose
Unit SizeEach
Contents & Storage
Storage conditions: 2°C to 8°C. Store in cool, dry conditions in well-sealed containers.