SST Sample Loops for Split-Loop Autosamplers
SST Sample Loops for Split-Loop Autosamplers
Actual product may vary
Thermo Scientific™
SST Sample Loops for Split-Loop Autosamplers
Order the Thermo Scientific™ SST Sample Loop you need for use in Thermo Scientific™ Dionex™ UltiMate™ split-loop autosamplers. Available in sizes from 25 to 1000μL.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSample Capacity
6820.2454500 μL
6820.24291000 μL
6820.2451100 μL
6820.245225 μL
6820.2453250 μL
Catalog number 6820.2454
Price (INR)
-
Request A Quote
Sample Capacity:
500 μL
Order the Thermo Scientific™ SST Sample Loop you need for use in Thermo Scientific™ Dionex™ UltiMate™ split-loop autosamplers. Available in sizes from 25 to 1000μL, the stainless steel (SST) sample loops incorporate Thermo Scientific™ Dionex™ Viper™ Fingertight Fittings which eliminate dead volumes and ensure easy, perfect connections in your HPLC or UHPLC system every time.
Specifications
For Use With (Equipment)Dionex UltiMate 3000 Split-Loop Autosampler WPS-3000(T)RS, Dionex UltiMate 3000 Split-Loop Autosampler WPS-3000(T)SL
MaterialStainless Steel
Sample Capacity500 μL
TypeSample Loop
Unit SizeEach