Sample Loops for UltiMate™ WPS-3000T PL RS Pulled Loop Autosamplers
Sample Loops for UltiMate™ WPS-3000T PL RS Pulled Loop Autosamplers
Actual product may vary
Thermo Scientific™
Sample Loops for UltiMate™ WPS-3000T PL RS Pulled Loop Autosamplers
Order the sample loop (1-125μL) you need for the Thermo Scientific™ Dionex™ UltiMate™ WPS-3000T PL RS pulled loop autosampler.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSample Capacity
6826.245050 μL
6826.24011 μL
6826.24055 μL
6826.242020 μL
6826.2412125 μL
6826.241010 μL
Catalog number 6826.2450
Price (INR)
-
Request A Quote
Sample Capacity:
50 μL
Order the Thermo Scientific™ Dionex™ Sample Loops (1-125μL) you need for the Thermo Scientific™ Dionex™ UltiMate™ WPS-3000T PL RS pulled loop autosampler. The sample loops incorporate Thermo Scientific™ Dionex™ nanoViper™ Fingertight Fittings which eliminate dead volumes and ensure easy, perfect connections in your HPLC or UHPLC system every time.
nanoViper is the new, fingertight connection system for nano LC connections which eliminates the assembly of PEEK™ sleeve connections. It is preassembled to maximize ease-of-use, and is fingertight, making over tightening impossible. The nanoViper fitting is capable of withstanding pressures up to a 1000 bar and is compatible with third party valves and unions.
Specifications
For Use With (Equipment)Dionex UltiMate 3000 Pulled Loop Autosampler WPS-3000(T)PL RS
Max. Pressure1000 bar
Sample Capacity50 μL
TypeSample Loop, NanoViper
Unit SizeEach