Viper™ Capillary Kit for Biocompatible RSLC Systems
Viper™ Capillary Kit for Biocompatible RSLC Systems
Thermo Scientific™
Viper™ Capillary Kit for Biocompatible RSLC Systems
Eliminate dead volumes and make perfect connections every time in your biocompatible LC systems with this fingertight fitting kit.
Have Questions?
Catalog NumberFor Use With (Equipment)
6841.2301Dionex UltiMate 3000 Biocompatible RSLC Systems
Catalog number 6841.2301
Price (INR)
-
Request A Quote
For Use With (Equipment):
Dionex UltiMate 3000 Biocompatible RSLC Systems
Avoid data-destroying dead volumes in the capillary connections of your biocompatible HPLC or UHPLC system by using the Thermo Scientific™ Viper™ Capillary Kit for Biocompatible RSLC Systems. Even with conventional columns and moderate pressures, poor connections can cause retention time shifts and peak distortions. Thermo Scientific™ Viper™ Fingertight Fittings which seal at the tip of the tubing and eliminate dead volumes and make it easier to plumb your HPLC or UHPLC system.
See Viper and nanoViper resources
Specifications
DescriptionViper Capillary Kit
For Use With (Equipment)Dionex UltiMate 3000 Biocompatible RSLC Systems
Product LineViper
TypeCapillary Kit
Unit SizeEach