UltiMate™ 3000 Split Loop Autosamplers Injection Volume Kits
UltiMate™ 3000 Split Loop Autosamplers Injection Volume Kits
Actual product may vary
Thermo Scientific™
UltiMate™ 3000 Split Loop Autosamplers Injection Volume Kits
Select an injection volume kit for your split-loop autosampler.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Sample Capacity
6841.2443Ultimate 3000 Split-Loop Autosamplers500 μL
6822.2436WPS-3000(T)SL Semi Prep1000 μL
6822.2442WPS-3000(T)RS, WPS-3000(T)SL250 μL
6841.2442Ultimate 3000 Split-Loop Autosamplers250 μL
6822.2443WPS-3000(T)RS, WPS-3000(T)SL500 μL
Catalog number 6841.2443
Price (INR)
-
Request A Quote
For Use With (Equipment):
Ultimate 3000 Split-Loop Autosamplers
Sample Capacity:
500 μL
Change or order new injection loops when your applications demand. Thermo Scientific™ Dionex™ UltiMate™ 3000 Split Loop Autosampler Injection Volume Kits. They are available in 250, 500 and 1000μL and include Thermo Scientific™ Dionex™ Viper™ Fingertight Fittings to ensure dead-volume-free connections.
Specifications
For Use With (Equipment)Ultimate 3000 Split-Loop Autosamplers
Sample Capacity500 μL
TypeInjection Volume Kit
Unit SizeEach