MP35N Biocompatible Sample Loops for Split-Loop Autosamplers
Product Image
Thermo Scientific™

MP35N Biocompatible Sample Loops for Split-Loop Autosamplers

Order the MP35N biocompatible sample loop (25-500μL) you need for use in Thermo Scientific™ Dionex™ UltiMate™ split-loop autosamplers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSample Capacity
6841.2454500 μL
6841.2451100 μL
6841.245225 μL
6841.2453250 μL
Catalog number 6841.2454
Price (INR)
-
Request A Quote
Sample Capacity:
500 μL
Order the Thermo Scientific™ MP35N Biocompatible Sample Loop you need for use in Thermo Scientific™ Dionex™ UltiMate™ split-loop autosamplers. Available in sizes from 25 to 500μL and made of a nickel alloy, the sample loops incorporate Thermo Scientific™ Dionex™ Viper™ Fingertight Fittings which eliminate dead volumes and ensure easy, perfect connections in your HPLC or UHPLC system every time.
Specifications
Biocompatible VersionYes
For Use With (Equipment)Dionex UltiMate 3000 Split-Loop Autosampler
MaterialMP35N
Sample Capacity500 μL
TypeSample Loop
Unit SizeEach