MP35N Biocompatible Buffer Loop for Split Loop Autosamplers
Product Image
Thermo Scientific™

MP35N Biocompatible Buffer Loop for Split Loop Autosamplers

Order biocompatible buffer loops for Thermo Scientific™ Dionex™ Split-loop Autosamplers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSample Capacity
6841.2468>250 μL
6841.2466100 μL
Catalog number 6841.2468
Price (INR)
-
Request A Quote
Sample Capacity:
>250 μL

Order the Thermo Scientific™ Dionex™ MP35N Biocompatible Buffer Loops (100 and greater than 250μL) you need for Thermo Scientific™ Dionex™ UltiMate™ split loop autosamplers. The buffer loops incorporate Thermo Scientific™ Dionex™ Viper™ Fingertight Fittings which eliminate dead volumes and ensure easy, perfect connections in your HPLC or UHPLC system every time.

Specifications
Biocompatible VersionYes
For Use With (Equipment)Dionex UltiMate 3000 Split-Loop Autosamplers
MaterialMP35N
Sample Capacity>250 μL
TypeBuffer Loop
Unit SizeEach