Vanquish™ Split Sampler Sample Loops
Vanquish™ Split Sampler Sample Loops
Actual product may vary
Thermo Scientific™
Vanquish™ Split Sampler Sample Loops
Thermo Scientific™ Vanquish™ Split Sampler Sample Loops come in a variety of sizes to fit your application requirements.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Sample CapacityBiocompatible Version
6850.1915Vanquish Split Sampler HT and FT10 μLYes
6850.1911Vanquish Split Sampler HT and FT25 μLYes
6850.1913Vanquish Split Sampler HT and FT100 μLYes
6851.1980Vanquish Split Sampler C/CT1000 μLNo
6851.1970Vanquish Split Sampler C/CT250 μLNo
6851.1960Vanquish Split Sampler C/CT10 μLNo
6851.1950Vanquish Split Sampler C/CT100 μLNo
6851.1940Vanquish Split Sampler C/CT25 μLNo
6252.1940Vanquish Split Sampler NT25 μLYes
6252.1950Vanquish Split Sampler NT100 μLYes
6252.1960Vanquish Split Sampler NT10 μLYes
Catalog number 6850.1915
Price (INR)
-
Request A Quote
For Use With (Equipment):
Vanquish Split Sampler HT and FT
Sample Capacity:
10 μL
Biocompatible Version:
Yes
Automate sample injection into your Thermo Scientific™ Vanquish™ HPLC or UHPLC system with Thermo Scientific™ Vanquish™ Split Sampler Sample Loops. These sample loops are available in 10, 25, 100, 250, 1000 μL sizes to meet your application requirements. Thermo Scientific™ Viper™ Fittings are included with our sample loops to make them easy to install with leak-free connections.
Vanquish Split Sampler Sample Loops use Viper Fittings, which have a ferrule-free tightening design, for virtually zero-dead-volume connections.
Specifications
Biocompatible VersionYes
For Use With (Equipment)Vanquish Split Sampler HT and FT
Sample Capacity10 μL
TypeSample Loop
Unit SizeEach