Refrigerator and Freezer Alarms
Thermo Scientific™
Refrigerator and Freezer Alarms
Meet specific application needs with these refrigerator and freezer alarms, offered in a variety of styles.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
6903Alarm Delay Module
6224Deluxe Remote Alarm
6714Electronic Local Alarm (for Freezers)
Catalog number 6903
Price (INR)
-
Type:
Alarm Delay Module
Meet specific application needs with Thermo Scientific™ Refrigerator and Freezer Alarms, optional alarms for lab refrigerators and freezers. These alarms are available in a variety of styles.
Alarm Delay Module
  • Eliminates nuisance alarms due to intermittent voltage flux from lightning or brief power outages
  • Adjustable delay releases signal to alarm/monitoring system only when alarm condition continues for specified time period
  • Digital timer can be set from 0.5 to 32 minutes
  • Red alarm light flashes to indicate transient event; must be manually reset
  • Signals chronic alarm conditions that may indicate impending component failure; allows time for inspection or corrective maintenance
  • Inserts variable time interval between detection of alarm condition and transmission of signal
  • Includes accurate digital timing circuit adjustable from 0.5, 1, 2, 4, 8, 18 or 32 minutes
  • Operates independent of line power; built-in battery and alarm test uses customer-supplied 9V battery
  • Overall dimensions, 1.62LX2.5WX4.75in.H (1.6X6.4X12.1cm)
Deluxe Electronic Remote Alarm System
  • Can be programmed to sound an alarm in the event of a temperature increase or power failure. 120V/60Hz
  • Alarm can dial up to four telephone numbers
  • Signals an alarm condition across any telephone system that accepts pulse dialing
  • One system can monitor up to four individual freezers or up to three groups of freezers
  • Requires dedicated phone line
  • Not for sale in countries that require a CE mark
Electronic Local Alarm Systems
  • Use with high-performance lab refrigerators and freezers
  • Freestanding system includes keyed ON/OFF alarm switch, audible and visual alarms for power failure and temperature deviation, audio silence button, and alarm test button
  • Painted metal cabinet with digital temperature display and keyed alarm switch
  • Must be customer installed
Standard Remote Alarm System
  • Use with general-purpose freezers and refrigerators; chromatography refrigerators equipped with accessory electronic local alarm system; and ultralow-temperature freezers
  • Can be located up to one-third mile from unit and used with either normally open or normally closed contacts
  • Emits both audible and visual signal in the event of power failure or temperature deviation
  • Wire not included
  • For 115V
  • Requires factory-installed alarm system
Specifications
Cabinet MaterialMetal
DefrostManual
Exterior FinishPainted
For Use With (Equipment)Thermo Scientific™ Refrigerators and Freezers
Monitoring OptionsAlarm
TypeAlarm Delay Module
WarrantyWarranty varies by country, please contact us for details
Certifications/ComplianceCE
StyleBuilt-in
Voltage120 V
Unit SizeEach