Search
| Catalog Number | Material | Septum | Color (Seal) | Vial Type |
|---|---|---|---|---|
| 6AK195W also known as SURESTART | Clear Glass | Silicone/PTFE, Pre-slit | White/Blue | SureStop Screw Thread Vial (Assembled) |
| 6AK592W | Clear Glass | Silicone/PTFE | White/Red | SureStop Short Thread Vial |
| 6AKSV9PP-15PI | Polypropylene | Silicone/Polyamide | White/Red | Short Thread Vial |
| 6AK594W | Amber Glass | Silicone/PTFE | White/Red | SureStop Short Thread Vial |
| 6AK599W | Clear Glass | Silicone/PTFE | Blue/Clear | Short Thread Vial |
| 6AK78BW | Clear Glass | Silicone/PTFE, Bonded | White/Red | Short Thread Vial |
| 6AK80W | Clear Glass | Silicone/PTFE | Red/Ivory | Short Thread Vial |
| 6AK82W | Amber Glass | Silicone/PTFE | Red/Ivory | Short Thread Vial |
| 6AK86W | Clear Glass | PTFE/Silicone/PTFE | Red/White/Red | Short Thread Vial |
| 6AK92W | Clear Glass | Silicone/PTFE | White/Red | Short Thread Vial |
| 6AK93BW | Clear Glass | Silicone/PTFE, Pre-slit, Bonded | White/Red | Short Thread Vial |
| 6AK94W | Amber Glass | Silicone/PTFE | White/Red | Short Thread Vial |
| 6AK95W | Clear Glass | Silicone/PTFE, Pre-slit | Blue/Clear | Short Thread Vial |
| 6AK186W also known as SURESTART | Clear Glass | PTFE/Silicone/PTFE | Red/White/Red | SureStop Screw Thread Vial (Assembled) |
| 6AK180W also known as SURESTART | Clear Glass | Silicone/PTFE | Red/Ivory | SureStop Screw Thread Vial (Assembled) |
| 6AK192W also known as SURESTART | Clear Glass | Silicone/PTFE | White/Red | SureStop Screw Thread Vial |
| 6AK595W also known as SURESTART | Clear Glass | Silicone/PTFE, Pre-slit | White/Blue | SureStop Screw Thread Vial |
| 6AK188W | Hydrolytic Glass (Class I) | PTFE/Silicone/PTFE | Red/White/Red | Screw Thread Vial |
| 6AK196W | Amber Hydrolytic Glass (Class I) | Silicone/PTFE, Pre-slit | White/Blue | Screw Thread Vial |
Specification Certified Kits
For everyday analysisAll SureSTART products are certified to meet physical parameter specifications as part of our manufacturing process.
SureStop Vials and Advanced Vial Closure System (AVCS) Caps: This patented screw top vial/cap combination features a push-through resistant sealing ring design, controlled septum compression, and SureStop closure positioning flange to ensure an effective seal that eliminates under- and over-sealing, reduces solvent evaporation, and creates less autosampler interruptions. SureStop vials and AVCS caps have universal instrument compatibility for all LC and GC autosamplers. They also contain low leachable levels, making them an ideal alternative to bonded caps for those seeking cleaner spectra.
The SureSTART Performance Levels
SureSTART vials, caps, inserts, septa, and kits are available in three performance levels. Now you can choose the products that best suit your analytical, performance, or budget needs.
Level 1: Everyday analysis
Chromatography approvedSureSTART Level 1 products are cost optimized, suitable for all instrument types, and quality tested to deliver the performance, sample security, and reproducibility you need for everyday analyses. They are an ideal solution for verifying compound presence and quantification using detection techniques such as UV, RI, and FID.
Level 2: High-throughput analysis
When robustness and reproducibility are key
SureSTART Level 2 vials have very low metal contents and leachables to ensure that they conform to the high standards you need to meet your regulatory requirements. The septa are conditioned to provide a very low level of chromatographic extractables. These products support routine analyses and deliver high quality data with an uninterrupted workflow.Level 3: High performance
When sensitivity is a mustSureSTART Level 3 products are packed in a clean environment according to our own hygiene concept and regulations to protect against contaminating factors. The vials contain the lowest levels of extractable metals and leachables to ensure consistent, reproducible sensitivity and selectivity for the most challenging analytes. The septa are conditioned to provide the lowest level of chromatographic extractables. Choose these products for ultimate method performance and high levels of and sample security and integrity.
All SureSTART vials, closures, and septa are certified to meet physical parameter specifications as part of our manufacturing process.