SureSTART™ GOLD-Grade Polypropylene Microvials for <2 mL Samples (Level 3)
Thermo Scientific™

SureSTART™ GOLD-Grade Polypropylene Microvials for <2 mL Samples (Level 3)

Use SureSTART™ GOLD-Grade Polypropylene Microvials, Performance Level 3, for analysis of <2 mL samples that are prone to non-specific binding
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeVolume (Metric) TotalFor Use With (Equipment)
6PSV9-04GPPScrew Thread Microvial0.4 mL9 mm Screw Caps and Septa
6PRV11-08GPPCSnap Microvial0.8 mL11 mm Snap Caps and Septa
6PRV11-03GPPCSnap Microvial0.3 mL11 mm Snap Caps and Septa
Catalog number 6PSV9-04GPP
Price (INR)
-
Product Type:
Screw Thread Microvial
Volume (Metric) Total:
0.4 mL
For Use With (Equipment):
9 mm Screw Caps and Septa

Use Performance Level 3 Thermo Scientific™ SureSTART™ 2 mL GOLD-Grade Polypropylene Microvials for <2 mL samples that are prone to non-specific binding. These low-bind polypropylene vials have been coated with a 200 nm thick layer of 100% silicon dioxide providing superior chemical inertness and performance. The surface is free from anions and cations, minimizing adsorption and ensuring that even trace-level samples are recovered reliably.

They can be used with all HPLC/UHPLC instrument types. These clear plastic 2 mL screw top vials are Specification Certified to ensure autosampler compatible physical dimensions. Seal these screw top vials with 9 mm screw caps and septa, which are sold separately

The SureSTART Performance Levels

SureSTART vials, caps, inserts, septa, and kits are available in three performance levels. Now you can choose the products that best suit your analytical, performance, or budget needs.

Level 1 – Everyday analysis

Chromatography approved SureSTART Level 1 products are cost optimized, suitable for all instrument types, and quality tested to deliver the performance, sample security, and reproducibility you need for everyday analyses. They are an ideal solution for verifying compound presence and quantification using detection techniques such as UV, RI, and FID.

Level 2 – High-throughput analysis

When robustness and reproducibility are key SureSTART Level 2 vials have very low metal contents and leachables to ensure that they conform to the high standards you need to meet your regulatory requirements. The septa are conditioned to provide a very low level of chromatographic extractables. These products support routine analyses and deliver high quality data with an uninterrupted workflow.

Level 3 – High performance

When sensitivity is a must SureSTART Level 3 products are packed in a clean environment according to our own hygiene concept and regulations to protect against contaminating factors. The vials contain the lowest levels of extractable metals and leachables to ensure consistent, reproducible sensitivity and selectivity for the most challenging analytes. The septa are conditioned to provide the lowest level of chromatographic extractables. Choose these products for ultimate method performance and high levels of and sample security and integrity.

All SureSTART vials, closures, and septa are certified to meet physical parameter specifications as part of our manufacturing process.

Specifications
Base StyleInsert Vial, Conical
Certifications/ComplianceSpecification Certified
For Use With (Equipment)9 mm Screw Caps and Septa
Height (Metric)32 mm
LevelLevel 3
Product TypeScrew Thread Microvial
Size (Metric) Opening9 mm
Volume (Metric) Residual4 μL
Volume (Metric) Total0.4 mL
Volume (Metric) Usable0.3 mL
ColorClear
Marking SpotNo
MaterialPolypropylene
Product LineSureSTART Level 3
Volume (Metric)2 mL
Unit SizePack of 100

Frequently asked questions (FAQs)

What kinds of applications or analytes are GOLD-Grade polypropylene vials particularly suited for?

Given their design features, GOLD-Grade Polypropylene vials are especially suited for:
• Trace-level LC/MS or GC/MS workflows where adsorption or loss of analyte could impact sensitivity.
• Small-volume samples (<1 mL) where residual volume needs to be minimal and sample recovery high.
• Samples prone to non-specific binding - e.g., peptides, proteins, bio-molecules, highly polar compounds, sticky analytes.
• High-sensitivity or regulated workflows (bio-pharma, metabolomics, environmental trace analysis) where sample integrity and low background are critical.

What performance levels are offered in the SureSTART collection, and where do SureSTART GOLD-Grade polypropylene vials fit?

The SureSTART collection categorizes products into three performance levels:
• Level 1—Everyday analysis: cost-optimized for general chromatography, basic detection.
• Level 2—High throughput: enhanced robustness, reproducibility, for regulated labs, MS workflows.
• Level 3—High performance: lowest extractables/leachables, packed in a clean environment, for the most demanding analytes (low-level, high-sensitivity) and HRAM-MS workflows.
The “GOLD-Grade” Level 3 vials provide the ultimate performance.
GOLD-Grade polypropylene vials are Level 3, meaning they are intended for high-sensitivity analysis where sample integrity matters most.

What sample volumes and formats do SureSTART GOLD-Grade polypropylene vials support?

SureSTART GOLD-Grade polypropylene vials support <1 mL samples. They feature fixed inserts allowing for a total volume of 0.3 mL, 0.4 mL, and 0.8 mL. They are available in either 9 mm screw thread or 11mm snap cap formats. The vial format is designed for samples that are small, precious, or highly concentrated.

How do SureSTART GOLD-Grade polypropylene vials differ from standard polypropylene autosampler vials?

Unlike standard vials, SureSTART GOLD-Grade polypropylene vials include:
• A pure silicon dioxide coating on the internal surface provides a low-bind coating, significantly reducing adsorption and ionic interactions.
• They are classified as Performance Level 3 (“High performance”) in the SureSTART collection, meaning they are manufactured in clean environments, have the lowest levels of extractables/leachables, and are intended for the most demanding analyses.
• They are labeled “GOLD-Grade” because of this special surface treatment.

What specific use case do SureSTART GOLD-Grade polypropylene vials help address?

These low-bind polypropylene screw- or snap-top vials are designed for samples of < 2 mL that are prone to non-specific binding, adsorption, pH shifts, or trace-level loss. These GOLD-Grade vials have been coated with a layer of 100% silicon dioxide, providing superior chemical inertness and performance, meaning they address challenging sample types (very low concentration, sticky analytes, adsorption-prone compounds) where standard vials might cause sample loss or signal suppression.