Corona™ CAD Inlet Gas Conditioning Module
Product Image
Thermo Scientific™
Corona™ CAD Inlet Gas Conditioning Module
Protect your charged aerosol detector from moisture and potentially damaging particles in your purified nitrogen gas.
Have Questions?
Catalog NumberFor Use With (Equipment)
70-8285Dionex Corona CAD Charged Aerosol Detector
Catalog number 70-8285
Price (INR)
-
Request A Quote
For Use With (Equipment):
Dionex Corona CAD Charged Aerosol Detector
Avoid moisture and potentially damaging particles in your purified nitrogen gas. The Thermo Scientific™ Dionex™ Corona™ CAD Inlet Gas Conditioning Module removes particles (0.1μm) and water to protect your charged aerosol detector and ensure the quality of your liquid chromatography results.
Specifications
Certifications/ComplianceCE marked,UL listed, CSA, FCC
Detection ModeCharged Aerosol Detection
For Use With (Equipment)Dionex Corona CAD Charged Aerosol Detector
TypeInlet Gas Conditioning Module
Unit SizeEach