Product Image
Thermo Scientific™
Rheodyne™ 7010 Sample Injector
Provides accurate and precise high-volume injections.
Have Questions?
Catalog NumberMode
7010TS
also known as 7010
Single Mode
Catalog number 7010TS
also known as 7010
Price (INR)
-
Mode:
Single Mode
Ensure accurate and precise high-volume HPLC injections of 5μL to 20mL samples with the Thermo Scientific™ Rheodyne™ 7010 Sample Injector. This single-mode injector uses the complete filling method to load the sample loop.
Specifications
DescriptionComplete Filling Method
Model7010
ModeSingle Mode
No. of Ports6 Ports
Stator MaterialStainless Steel
TypeSample Injector
Volume (Metric) Sample5 μL to 5 mL
Unit SizeEach