Refrigerator and Freezer Door Options
Thermo Scientific™
Refrigerator and Freezer Door Options
Customize refrigerator and freezer configurations to better suit the needs of the lab with these refrigerator and freezer door options.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)TypeHinge Side
701445.8 cu. ft. UnitsRefrigerator and Freezer Door, GlassLeft/Right
699511.5 cu. ft. UnitsRefrigerator and Freezer Door, SolidLeft
6996TS
also known as 6996
11.5 cu. ft. UnitsRefrigerator and Freezer Door, GlassLeft
699723.3 cu. ft. UnitsRefrigerator and Freezer Door, SolidLeft
699823.3 cu. ft. UnitsRefrigerator and Freezer Door, GlassLeft
6999TS
also known as 6999
29.2 cu. ft. UnitsRefrigerator and Freezer Door, SolidLeft
700145.8 cu. ft. UnitsRefrigerator and Freezer Door, SolidLeft/Left
7012TS
also known as 7012
23.3 cu. ft. UnitsRefrigerator and Freezer Door, GlassRight
706611.5 cu. ft. UnitsRefrigerator and Freezer Door, GlassRight
706745.8 cu. ft. UnitsRefrigerator and Freezer Door, GlassLeft/Left
706845.8 cu. ft. UnitsRefrigerator and Freezer Door, SolidRight/Right
706945.8 cu. ft. UnitsRefrigerator and Freezer Door, GlassRight/Right
707311.5 cu. ft. UnitsRefrigerator and Freezer Door, Heated GlassLeft
707411.5 cu. ft. UnitsRefrigerator and Freezer Door, Heated GlassRight
7075TS
also known as 7075
23.3 cu. ft. UnitsRefrigerator and Freezer Door, Heated GlassLeft
708623.3 cu. ft. UnitsRefrigerator and Freezer Door, Heated GlassRight
708729.2 cu. ft. UnitsRefrigerator and Freezer Door, Heated GlassLeft
708829.2 cu. ft. UnitsRefrigerator and Freezer Door, Heated GlassRight
708945.8 cu. ft. UnitsRefrigerator and Freezer Door, Heated GlassLeft/Left
709045.8 cu. ft. UnitsRefrigerator and Freezer Door, Heated GlassRight/Right
709145.8 cu. ft. UnitsRefrigerator and Freezer Door, Heated GlassLeft/Right
71184.7 cu. ft. Forma, Jewett, Puffer HubbardRefrigerator and Freezer Door, GlassRight
71194.7 cu. ft. Forma, Jewett, Puffer HubbardRefrigerator and Freezer Door, SolidLeft
71234.7 cu. ft. UnitsRefrigerator and Freezer Door, GlassLeft
LE701329.2 cu. ft. UnitsRefrigerator and Freezer Door, GlassRight
Catalog number 7014
Price (INR)
-
For Use With (Equipment):
45.8 cu. ft. Units
Type:
Refrigerator and Freezer Door, Glass
Hinge Side:
Left/Right
Customize refrigerator and freezer configurations to better suit the needs of the lab with Thermo Scientific™ Refrigerator and Freezer Door Options which include glass and heated glass doors, as well as either right- or left-hand swing.
For use with Thermo Scientific™ Forma™, Jewett™, Puffer Hubbard™ and Revco™ series. Solid doors are standard on most refrigerators unless otherwise specified. Optional dual-pane glass doors must be specified when ordering. Contact your Sales Representative for information about heated glass doors.
  • Glass doors standard on selected blood bank, chromatography and pharmacy refrigerators
  • Sliding glass doors standard on all 45.8 cu. ft. (1297L) models
  • Heated glass doors increase amperage and may require next size breaker
Ordering Information:
Door options must be specified at time of order
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Door MaterialGlass (Door)
For Use With (Equipment)45.8 cu. ft. Units
Hinge SideLeft/Right
TypeRefrigerator and Freezer Door, Glass
Capacity (English)51.1 cu. ft.
Capacity (Metric)1447 L
Door StyleHinged
Unit SizeEach