4-column drawer divider for 4.9 and 11.5 cu. ft. units
Thermo Scientific™
4-column drawer divider for 4.9 and 11.5 cu. ft. units
Maximize storage inside blood bank refrigerators and plasma freezers with these optional drawer dividers.
Have Questions?
Catalog number 7027
Price (INR)
-
Maximize storage inside blood bank refrigerators and plasma freezers with these optional Thermo Scientific™ Drawer Dividers.
Stainless-steel drawer dividers help manage inventory of bags and boxes:
  • Four column dividers for all models except 29.2 cu. ft. (826L) models
  • Five column dividers for 29.2 cu. ft. (826L) models
Specifications
Description4-column divider
HoldsBags
For Use With (Equipment)4.9 and 11.5 cu. ft. Thermo Scientific Revco and Forma blood bank refrigerators and plasma freezers.
Type4-Column Divider
Unit SizeEach