CaptureSelect™ Biotin Anti-EPO Conjugate
Thermo Scientific™

CaptureSelect™ Biotin Anti-EPO Conjugate

Thermo Scientific™ CaptureSelect™ Biotin Anti-EPO Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) with high affinity and selectivity for both recombinant and natural-sourced human erythropoietin (EPO).
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
7103372500500 μg
7103372100100 μg
Catalog number 7103372500
Price (INR)
-
Quantity:
500 μg

CaptureSelect Biotin Anti-EPO Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) with high affinity and selectivity for both recombinant and natural-sourced human erythropoietin (EPO). The affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces.

Applications for CaptureSelect Biotin Anti-EPO Conjugate include ELISA, western blot, Gyrolab™-based immunoassays, and use of label-free detection platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).

Animal origin statement

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Ligand TypeAntibody
Product LineCaptureSelect
Product TypeLigand Conjugates
Quantity500 μg
Shipping ConditionApproved for shipment on Wet or Dry Ice
Label TypeBiotin and Other Haptens
Target SpeciesHuman
Unit SizeEach
Contents & Storage
Store at -5°C to -30°C.