CaptureSelect™ Biotin Anti-VSVG Conjugate
Thermo Scientific™
CaptureSelect™ Biotin Anti-VSVG Conjugate
CaptureSelect Biotin Anti-VSVG Conjugate consists of a 13-kDa recombinant single domain antibody fragment (VHH affinity ligand) that binds with high affinity and selectivity to the ectodomain of VSVG (glycoprotein (G) of vesicular stomatitis virus).
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
7104082500500 μg
7104082100100 μg
Catalog number 7104082500
Price (INR)
227,976.00
500 μL
Quantity:
500 μg
Price (INR)
227,976.00
500 μL
CaptureSelect Biotin Anti-VSVG Conjugate consists of a 13-kDa recombinant single domain antibody fragment (VHH affinity ligand) that binds with high affinity and selectivity to the ectodomain of VSVG (glycoprotein (G) of vesicular stomatitis virus).
The affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces. This conjugate enables detection and quantitation of free VSVG protein, membrane bound VSVG as displayed on vesicular stomatitis virus, and VSVG pseudo-typed viruses and extracellular vesicles (EVs).
Applications for CaptureSelect Biotin Anti-VSVG Conjugate include ELISA, Gyrolab™-based immunoassays, immunoprecipitation with streptavidin-coupled Dynabeads, and label-free detection platforms such as those based on surface plasmon resonance (SPR) and bio-layer interferometry (BLI).
To set up a sensitive sandwich ELISA for quantitation of free VSVG protein and VSVG-bearing particles, we recommend the use of CaptureSelect Biotin Anti-VSVG Conjugate in combination with CaptureSelect HRP Anti-VSVG Conjugate (Cat. No. 7304072100) as detection reagent .
Animal origin statement
CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration1 mg/mL
FormulationPBS (pH 7.4)
Quantity500 μg
Shipping ConditionDry ice
TargetVSV Glycoprotein
FormFrozen Liquid
Product LineCaptureSelect
TypeBiotin
Unit Size500 μL
Contents & Storage
Store in freezer (-30°C to -5°C).